5-Methoxysalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde

16416-0050 16416-0100 16416-0500
TS16416-0050EA 109 USD
TS16416-0050 TS16416-0100 TS16416-0500
5-Methoxysalicylaldehyde 98%
5-Methoxysalicylaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 103 °C (2.5 mmHg)
Melting Pt: 4 °C
Density: 1.21 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR