7-Amino-1,3-naphthalenedisulfonic acid tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
16396-0025
16396-1000
16396-5000
TS16396-0025EA
389
USD
TS16396-0025
TS16396-1000
TS16396-5000
7-Amino-1,3-naphthalenedisulfonic acid tech.
7-Amino-1,3-naphthalenedisulfonic acid
Formula:
C₁₀H₁₂N₂O₆S₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003991 CAS Number: 86-65-7 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | Off-white to green-beige to light brown Powder or needles |
Infrared spectrum | Conforms |
Titration with NaOH | ≥60 % (As is) |
Water | ≤0.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...