3-Cyanobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Formylbenzonitrile

16390-0010 16390-0050 16390-0250
TS16390-0010EA 27 USD
TS16390-0010 TS16390-0050 TS16390-0250
3-Cyanobenzaldehyde 99%
3-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥98.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR