α-Bromo-m-toluonitrile 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile

16351-0100 16351-0500
TS16351-0100EA 86 USD
TS16351-0100 TS16351-0500
α-Bromo-m-toluonitrile 95%
α-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 130 °C (4 mmHg)
Melting Pt: 92…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00001809
CAS Number: 28188-41-2
EINECS: 248-890-1
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 97 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR