2-Chlorobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2-Chlorophenyl)methanol

16342-0100 16342-0500
TS16342-0100EA 30 USD
TS16342-0100 TS16342-0500
2-Chlorobenzyl alcohol 99%
2-Chlorobenzyl alcohol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 69…72 °C
MDL Number: MFCD00004604
CAS Number: 17849-38-6
EINECS: 241-801-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 69 to 72 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR