2,5-Dichlorobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

16324-0100 16324-0500 16324-2500
TS16324-0100EA 30 USD
TS16324-0100 TS16324-0500 TS16324-2500
2,5-Dichlorobenzoic acid 97%
2,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
Boiling Pt: 301 °C (760 mmHg)
Melting Pt: 152…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002416
CAS Number: 50-79-3
EINECS: 200-065-7

Order Now

Specification Test Results

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 152 to 157 °C
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR