2-Amino-p-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

16321-0500 16321-2500
TS16321-0500EA 31 USD
TS16321-0500 TS16321-2500
2-Amino-p-cresol 97%
2-Amino-p-cresol
Formula: C₇H₉NO
Melting Pt: 134…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00007699
CAS Number: 95-84-1
EINECS: 202-457-3

Order Now

Specification Test Results

Appearance Beige to grey-brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 134 to 139 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR