4-Amino-m-cresol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-hydroxytoluene, 4-Amino-3-methylphenol, 1-Amino-4-hydroxy-2-methylbenzene, 4-Hydroxy-2-methylaniline

16320-0500 16320-2500
TS16320-0500EA 80 USD
TS16320-0500 TS16320-2500
4-Amino-m-cresol 99+%
4-Amino-m-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 176…180 °C
MDL Number: MFCD00007871
CAS Number: 2835-99-6
EINECS: 220-621-2

Order Now

Specification Test Results

Appearance Off-white to yellow to beige to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 176 to 180 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR