α-Bromo-o-toluonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
16313-0050
16313-0250
16313-1000
TS16313-0050EA
46
USD
TS16313-0050
TS16313-0250
TS16313-1000
α-Bromo-o-toluonitrile 98%
α-Bromo-o-toluonitrile
Formula:
C₈H₆BrN Boiling Pt: 152…155 °C (15 mmHg) Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001794 CAS Number: 22115-41-9 UN: 1759 ADR: 8,II |
Specification Test Results
Appearance | White to grey-brown Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 70 to 74 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...