4-Hydroxy-3-nitrobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals

16289-0050 16289-0250
TS16289-0050EA 59 USD
TS16289-0050 TS16289-0250
4-Hydroxy-3-nitrobenzaldehyde 97%
4-Hydroxy-3-nitrobenzaldehyde
Formula: C₇H₅NO₄
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00007117
CAS Number: 3011-34-5
EINECS: 221-141-6

Order Now

Specification Test Results

Appearance Yellow-brown to brown Powder
Infrared spectrum Conforms
Melting point 139 to 144 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR