3,5-Xylenol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Hydroxy-3-xylene, 3,5-Dimethylphenol, 5-Hydroxy-m-xylene, 1,3-Dimethyl-5-hydroxybenzene

16279-0025 16279-0050 16279-1000 16279-5000
TS16279-0025EA 582 USD
TS16279-0025 TS16279-0050 TS16279-1000 TS16279-5000
3,5-Xylenol 99+%
3,5-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 222 °C (760 mmHg)
Melting Pt: 63…66 °C
Density: 1.115 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00002307
CAS Number: 108-68-9
EINECS: 203-606-5
UN: 2928
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Specification Test Results

Appearance White to yellow to orange Crystalline solid or fused mass
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR