Homovanillic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-Hydroxy-3-methoxyphenyl)acetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid

16253-0010 16253-0050
TS16253-0010EA 75 USD
TS16253-0010 TS16253-0050
Homovanillic acid 98%
Homovanillic acid
Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 141…145 °C
MDL Number: MFCD00004350
CAS Number: 306-08-1
EINECS: 206-176-7
Merck Index: 13,04761

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 145 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR