4-Bromophenylhydrazinium chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

16231-0100 16231-0500
TS16231-0100EA 126 USD
TS16231-0100 TS16231-0500
4-Bromophenylhydrazinium chloride 99%
4-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
Melting Pt: 210…215 °C
MDL Number: MFCD00012941
CAS Number: 622-88-8
UN: 3261

Order Now

Specification Test Results

Appearance Beige to grey to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 210 to 215 °C (Dec)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR