5-Bromosalicylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-hydroxybenzoic acid

16214-0500 16214-2500
TS16214-0500EA 80 USD
TS16214-0500 TS16214-2500
5-Bromosalicylic acid 98%
5-Bromosalicylic acid
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 162…166 °C
MDL Number: MFCD00002455
CAS Number: 89-55-4
EINECS: 201-917-0

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 164 to 172 °C
Titration with NaOH 97.5 to 102.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR