4,4'-Dimethoxybenzophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(4-methoxyphenyl)methanone

16174-0250 16174-1000
TS16174-0250EA 91 USD
TS16174-0250 TS16174-1000
4,4'-Dimethoxybenzophenone 97%
4,4'-Dimethoxybenzophenone
Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Melting Pt: 141…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00008404
CAS Number: 90-96-0
EINECS: 202-028-0

Order Now

Specification Test Results

Appearance Yellow to light beige Powder
Infrared spectrum Conforms
Melting point 141 to 146 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR