2,6-Dichlorobenzamide 97%

Supplier: Thermo Scientific Chemicals

16169-0250 16169-1000
TS16169-0250EA 28 USD
TS16169-0250 TS16169-1000
2,6-Dichlorobenzamide 97%
2,6-Dichlorobenzamide
Formula: C₇H₅Cl₂NO
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 196…201 °C
MDL Number: MFCD00007975
CAS Number: 2008-58-4
EINECS: 217-918-4

Order Now

Specification Test Results

Appearance White to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 196 to 201 °C
GC ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR