(±)-trans-1,2-Cyclohexanediol 98%

Supplier: Thermo Scientific Chemicals

16165-0100 16165-0500
TS16165-0100EA 47 USD
TS16165-0100 TS16165-0500
(±)-trans-1,2-Cyclohexanediol 98%
(±)-trans-1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
Boiling Pt: 117 °C (17mbar)
Melting Pt: 100…104 °C
Flash Pt: 134 °C
MDL Number: MFCD00063611
CAS Number: 1460-57-7
EINECS: 215-956-6

Order Now

Specification Test Results

Appearance Off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 100 to 104 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR