Veratrole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether

16111-0010 16111-1000 16111-5000
TS16111-0010EA 129 USD
TS16111-0010 TS16111-1000 TS16111-5000
Veratrole 99+%
Veratrole
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.08 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008357
CAS Number: 91-16-7
EINECS: 202-045-3
Merck Index: 12,10089

Order Now

Specification Test Results

Appearance Clear liquid or solid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5360 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR