2,4-Dichlorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

16090-0010 16090-0050 16090-1000 16090-2500
TS16090-0010EA 154 USD
TS16090-0010 TS16090-0050 TS16090-1000 TS16090-2500
2,4-Dichlorobenzoic acid 98%
2,4-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: >200 °C (760 mmHg)
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00002414
CAS Number: 50-84-0
EINECS: 200-067-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
Water ≤0.5 % (K.F.)
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %
Color scale ≤15 APHA
Apparent equivalent weight 104.0 to 106.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR