4-Fluorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Fluorobenzonitrile

16081-0050 16081-0250
TS16081-0050EA 37 USD
TS16081-0050 TS16081-0250
4-Fluorobenzonitrile 99%
4-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 188 °C (750 mmHg)
Melting Pt: 33…36 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001812
CAS Number: 1194-02-1
EINECS: 214-784-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 33 to 36 °C
GC ≥98.5 %
Stabilizer 0.1 % 4-tert-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR