(4-Chlorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals

16072-0025 16072-0050 16072-1000 16072-5000
TS16072-0025EA 777 USD
TS16072-0025 TS16072-0050 TS16072-1000 TS16072-5000
(4-Chlorophenyl)acetic acid 98%
(4-Chlorophenyl)acetic acid
Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
Water ≤0.2 % (K.F.)
HPLC ≥97.5 %
Appearance of solution (5% soln. in CHCl3) clear
Titration with NaOH 97.5 to 101.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR