1,3,5-Trimethoxybenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

16055-0100 16055-0500 16055-2500
TS16055-0100EA 42 USD
TS16055-0100 TS16055-0500 TS16055-2500
1,3,5-Trimethoxybenzene 99%
1,3,5-Trimethoxybenzene
Formula: C₉H₁₂O₃
Boiling Pt: 255 °C (760 mmHg)
Melting Pt: 50…53 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008385
CAS Number: 621-23-8
EINECS: 210-673-4

Order Now

Specification Test Results

Appearance White to cream to yellow Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR