2,4,6-Trimethoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals

16053-0100 16053-0250 16053-2500
TS16053-0100EA 52 USD
TS16053-0100 TS16053-0250 TS16053-2500
2,4,6-Trimethoxybenzaldehyde 98%
2,4,6-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00003313
CAS Number: 830-79-5
EINECS: 212-598-2

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 120 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR