(4-Bromophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

16052-0250 16052-1000
TS16052-0250EA 64 USD
TS16052-0250 TS16052-1000
(4-Bromophenyl)acetic acid 99%
(4-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00004342
CAS Number: 1878-68-8
EINECS: 217-523-7

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 115 to 118 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR