2,4-Dimethylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

16032-0100 16032-0500
TS16032-0100EA 54 USD
TS16032-0100 TS16032-0500
2,4-Dimethylbenzoic acid 98%
2,4-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 267 °C (727 mmHg)
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002480
CAS Number: 611-01-8
EINECS: 210-246-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 127 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR