Methyl vanillate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-hydroxy-3-methoxybenzoate
16030-0050
16030-0250
16030-1000
TS16030-0050EA
37
USD
TS16030-0050
TS16030-0250
TS16030-1000
Methyl vanillate 99%
Methyl vanillate
Formula:
C₉H₁₀O₄ Boiling Pt: 285…287 °C (760 mmHg) Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008438 CAS Number: 3943-74-6 EINECS: 223-525-9 |
Specification Test Results
Appearance | Light yellow to beige Powder and/or granules |
Infrared spectrum | Conforms |
Melting point | 64 to 67 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...