6-Nitro-m-cresol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Methyl-2-nitrophenol, 1-Hydroxy-3-methyl-6-nitrobenzene
16015-0250
16015-1000
TS16015-0250EA
48
USD
TS16015-0250
TS16015-1000
6-Nitro-m-cresol 97%
6-Nitro-m-cresol
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Melting Pt: 53…56 °C Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007111 CAS Number: 700-38-9 EINECS: 211-843-0 UN: 2446 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow to yellow-green Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 53 to 56 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...