2,4-Di-tert-butylphenol 97%

Supplier: Thermo Scientific Chemicals
Warning

16012-0025 16012-1000 16012-5000
TS16012-0025EA 157 USD
TS16012-0025 TS16012-1000 TS16012-5000
2,4-Di-tert-butylphenol 97%
2,4-Di-tert-butylphenol
Formula: C₁₄H₂₂O
Boiling Pt: 264 °C (760 mmHg)
Melting Pt: 54…59 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008828
CAS Number: 96-76-4
EINECS: 202-532-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
Melting point 54 to 59 °C
Water ≤0.1 % (K.F.)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR