Benzyl(dimethyl)amine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

15975-0025 15975-0051 15975-1000 15975-5000
TS15975-0025EA 242 USD
TS15975-0025 TS15975-0051 TS15975-1000 TS15975-5000
Benzyl(dimethyl)amine 99%
Benzyl(dimethyl)amine
Formula: C₆H₅CH₂N(CH₃)₂
MW: 135.21 g/mol
Boiling Pt: 183…184 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.9 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008329
CAS Number: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=7044105a7e7fbce7e00f1d15d7a4e017a1b9d418&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5020 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %
Filt.over 0.2 micron PFTE Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR