4-Methoxybenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol

15973-0025 15973-1000 15973-5000
TS15973-0025EA 445 USD
TS15973-0025 TS15973-1000 TS15973-5000
4-Methoxybenzyl alcohol 98%
4-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 259 °C (760 mmHg)
Melting Pt: 23…25.5 °C
Density: 1.1125 g/cm³
Flash Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00004653
CAS Number: 105-13-5
EINECS: 203-273-6
Merck Index: 13,00608

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5440 to 1.5470 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 1.111 to 1.115
Filt.over 0.2 micron PFTE Filtered
Acidity (CH3COOH) ≤0.0005 meq/g
Peroxides ≤0.0025 % (H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR