2-Chlorobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15924-0020
15924-0050
15924-1000
15924-5000
TS15924-0020EA
123
USD
TS15924-0020
TS15924-0050
TS15924-1000
TS15924-5000
2-Chlorobenzoic acid 98%
2-Chlorobenzoic acid
Formula:
C₇H₅ClO₂ Boiling Pt: 285 °C (760 mmHg) Melting Pt: 139…143 °C Flash Pt: 173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002412 CAS Number: 118-91-2 EINECS: 204-285-4 Merck Index: 13,02143 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 139 to 143 °C |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Copper (Cu) | ≤5 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤10 ppm |
Selenium (Se) | ≤10 ppm |
Zinc (Zn) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...