Trityl alcohol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Triphenylmethanol, Triphenylcarbinol

15891-0010 15891-0500 15891-2500
TS15891-0010EA 617 USD
TS15891-0010 TS15891-0500 TS15891-2500
Trityl alcohol 97%
Trityl alcohol
Formula: C₁₉H₁₆O
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00004445
CAS Number: 76-84-6
EINECS: 200-988-5
Merck Index: 13,09810

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
GC ≥96.0 %
Chloride (Cl) ≤140 ppm
Arsenic (As) ≤8 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR