Biphenyl-4-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [1,1'-Biphenyl]-4-ol, 4-Hydroxydiphenyl, 4-Phenylphenol, p-Hydroxybiphenyl, 4-Hydroxybiphenyl

15871-0010 15871-2500
TS15871-0010EA 198 USD
TS15871-0010 TS15871-2500
Biphenyl-4-ol 97%
Biphenyl-4-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: >163 °C
Flash Pt: 165 °C
MDL Number: MFCD00002347
CAS Number: 92-69-3
EINECS: 202-179-2
UN: 3077
Merck Index: 12,07459

Order Now

Specification Test Results

Appearance White to beige Flakes
Infrared spectrum Conforms
Melting point 163 to 168 °C
GC ≥96.0 %
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR