1-Amino-4-naphthol hydrochloride 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-1-naphthol hydrochloride

15851-0050 15851-0250
TS15851-0050EA 73 USD
TS15851-0050 TS15851-0250
1-Amino-4-naphthol hydrochloride 90%, Technical Grade
1-Amino-4-naphthol hydrochloride
Formula: C₁₀H₁₀ClNO
MW: 195.65
Melting Pt: 273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012562
CAS Number: 5959-56-8
EINECS: 227-727-8

Order Now

Specification Test Results

Appearance Pink to grey Powder
Infrared spectrum Conforms
HPLC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR