Aniline hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylamine HCl, Aminobenzene hydrochloride, Anilinium chloride, Aminobenzene HCl, Phenylamine hydrochloride

15841-0025 15841-1000 15841-5000
TS15841-0025EA 315 USD
TS15841-0025 TS15841-1000 TS15841-5000
Aniline hydrochloride 99%
Aniline hydrochloride
Formula: C₆H₈ClN
MW: 129.59 g/mol
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: 196…199 °C
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00012958
CAS Number: 142-04-1
EINECS: 205-519-8
UN: 1548
ADR: 6.1,III
Merck Index: 13,00616

Order Now

Specification Test Results

Appearance White to grey Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 196 to 200 °C
Titration with HClO4 ≥98.5 %
UV at 280 nm A: ≤0.05
Titration with NaOH ≥99.4 %
Gradient analysis passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR