Aniline 99.8%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aminobenzene, Phenylamine

15819-0010 15819-0025 15819-0050 15819-1000
TS15819-0010EA 128 USD
TS15819-0010 TS15819-0025 TS15819-0050 TS15819-1000
Aniline 99.8%, pure
Aniline
Formula: C₆H₅NH₂
MW: 93.13 g/mol
Boiling Pt: 181…185 °C (760 mmHg)
Melting Pt: –6.2 °C
Density: 1.0213 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00007629
CAS Number: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5875 (20°C, 589 nm)
GC ≥99.75 %
Sulfate (SO4) ≤0.03 %
Calcium (Ca) ≤0.005 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR