3,5-Dimethoxyphenol 97%

Supplier: Thermo Scientific Chemicals
Warning

15812-0100 15812-0500
TS15812-0100EA 97 USD
TS15812-0100 TS15812-0500
3,5-Dimethoxyphenol 97%
3,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 172…175 °C (17 mmHg)
Melting Pt: 40…45 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008388
CAS Number: 500-99-2
EINECS: 207-917-7

Order Now

Specification Test Results

Appearance White to yellow to pink Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 40 to 46 °C
GC ≥96.0 %
UV at 350 nm A: ≤0.01
pH 6.5 to 7.3 (5 % in water)
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR