4-Methoxybenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile

15805-0250 15805-1000
TS15805-0250EA 90 USD
TS15805-0250 TS15805-1000
4-Methoxybenzonitrile 99%
4-Methoxybenzonitrile
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 256…257 °C (765 mmHg)
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00001818
CAS Number: 874-90-8
EINECS: 212-871-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 60 °C
GC ≥98.5 %
UV at 300 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0001 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR