m-Toluidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Aminotoluene, 3-Toluidine, 3-Methylaniline
15787-0010
15787-0050
TS15787-0010EA
139
USD
TS15787-0010
TS15787-0050
m-Toluidine 99%
m-Toluidine
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 203 Melting Pt: -30 Density: 0.98 g/cm³ Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007808 CAS Number: 108-44-1 EINECS: 203-583-1 UN: 1708 ADR: 6.1 II Merck Index: 13,09614 |
Specification Test Results
Appearance | Clear colorless to yellow to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5655 to 1.5685 (20°C, 589 nm) |
GC | ≥98.5 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.002 % |
Sodium (Na) | ≤0.01 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...