1,2,4,5-Tetrachlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

15778-0050 15778-2500
TS15778-0050EA 24 USD
TS15778-0050 TS15778-2500
1,2,4,5-Tetrachlorobenzene 98%
1,2,4,5-Tetrachlorobenzene
Formula: C₆H₂Cl₄
MW: 215.89 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 139…142 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000549
CAS Number: 95-94-3
EINECS: 202-466-2

Order Now

Specification Test Results

Appearance Light pink to grey Crystalline solid
Infrared spectrum Conforms
Melting point 139 to 142 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR