5-Aminoindan 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Aminoindane

15735-0100 15735-0500
TS15735-0100EA 90 USD
TS15735-0100 TS15735-0500
5-Aminoindan 97%
5-Aminoindan
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 247…249 °C (745 mmHg)
Melting Pt: 34…36 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003803
CAS Number: 24425-40-9
EINECS: 246-241-7

Order Now

Specification Test Results

Appearance Brown Crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR