4-Nitrobenzaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15699-0250
15699-1000
15699-5000
TS15699-0250EA
56
USD
TS15699-0250
TS15699-1000
TS15699-5000
4-Nitrobenzaldehyde 99%
4-Nitrobenzaldehyde
Formula:
C₇H₅NO₃ MW: 151.12 g/mol Melting Pt: 105…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007346 CAS Number: 555-16-8 EINECS: 209-084-5 Merck Index: 12,06684 |
Specification Test Results
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 107 °C |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...