m-Anisaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

15676-0250 15676-1000 15676-5000
TS15676-0250EA 48 USD
TS15676-0250 TS15676-1000 TS15676-5000
m-Anisaldehyde 97%
m-Anisaldehyde
Formula: C₈H₈O₂
Boiling Pt: 143 °C (50 mmHg)
Density: 1.119 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1
EINECS: 209-712-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5550 (20°C, 589 nm)
GC ≥96.0 %
Chloride (Cl) ≤0.2 %
Iodide (I) (Limit about 0.005 %) Passes test
Bromate (BrO3) ≤0.002 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.002 %
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR