5-Iodovanillin 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

15667-0050 15667-0250
TS15667-0050EA 37 USD
TS15667-0050 TS15667-0250
5-Iodovanillin 97%
5-Iodovanillin
Formula: C₈H₇IO₃
MW: 278.05 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 180…184 °C
MDL Number: MFCD00006941
CAS Number: 5438-36-8
EINECS: 226-617-7

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 180 to 184 °C
HPLC ≥96.0 %
UV at 280 nm A: ≤0.01

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR