4-Iodoaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

15661-0250 15661-1000 156615000
TS15661-0250EA 56 USD
TS15661-0250 TS15661-1000 TS156615000
4-Iodoaniline 99%
4-Iodoaniline
Formula: C₆H₆IN
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 59…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007848
CAS Number: 540-37-4
EINECS: 208-743-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,05049

Order Now

Specification Test Results

Appearance Beige to grey-brown to dark grey Crystalline powder or needles
Infrared spectrum Conforms
Melting point 61 to 64 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR