4-Fluorobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

15615-0010 15615-0500 15615-2500
TS15615-0010EA 873 USD
TS15615-0010 TS15615-0500 TS15615-2500
4-Fluorobenzaldehyde 98+%
4-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 181 °C (758 mmHg)
Melting Pt: –10 °C
Density: 1.15 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003378
CAS Number: 459-57-4
EINECS: 207-293-6
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5195 to 1.5215 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR