Fluorene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9H-Fluorene, Diphenylenmethane

15613-0250 15613-1000 15613-5000
TS15613-0250EA 43 USD
TS15613-0250 TS15613-1000 TS15613-5000
Fluorene 98+%
Fluorene
Formula: C₁₃H₁₀
MW: 166.22 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 112…116 °C
Density: 1.2 g/cm³
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00001111
CAS Number: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9,III
Merck Index: 13,04181

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
Loss on drying ≤0.5 %
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR