3-Fluorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

15550-0010 15550-0050
TS15550-0010EA 29 USD
TS15550-0010 TS15550-0050
3-Fluorobenzylamine 97%
3-Fluorobenzylamine
Formula: C₇H₈FN
Boiling Pt: 87…89 °C (25 mmHg)
Density: 1.09 g/cm³
Flash Pt: 71 °C
MDL Number: MFCD00008113
CAS Number: 100-82-3
EINECS: 202-891-3
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5125 to 1.5175 (20°C, 589 nm)
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR