3-Methylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene

15548-0010 15548-0050 15548-0250
TS15548-0010EA 30 USD
TS15548-0010 TS15548-0050 TS15548-0250
3-Methylbenzylamine 98%
3-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 202…205 °C (760 mmHg)
Density: 0.966 g/cm³
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00008118
CAS Number: 100-81-2
EINECS: 202-890-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5380 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR