2,3-Dimethoxyphenol 99%

Supplier: Thermo Scientific Chemicals
Warning

15525-0050 15525-0250
TS15525-0050EA 79 USD
TS15525-0050 TS15525-0250
2,3-Dimethoxyphenol 99%
2,3-Dimethoxyphenol
Formula: C₈H₁₀O₃
Boiling Pt: 234 °C (1013 hPa)
Flash Pt: 108 °C
MDL Number: MFCD00008366
CAS Number: 5150-42-5
EINECS: 225-922-2

Order Now

Specification Test Results

Appearance Clear orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR